Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008325.4 | NP_032351.2 | 3 | 611 | ||
GenBank mRNA | AK031414.1 | - | 2 | 265 | ||
AK155627.1 | - | 3 | 612 | |||
AK169998.1 | - | 2 | 269 | |||
BC057935.1 | - | 2 | 232 | 5327150 | ||
BY749834.1 | - | 3 | 608 | |||
L34111.1 | - | 2 | 185 |