Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015176.3 | NP_055991.1 | 4 | 713 | ||
NR_049764.1 | 3 | 574 | ||||
GenBank mRNA | AB007952.1 | - | 4 | 464 | ||
AK001628.1 | - | 4 | 689 | |||
AK303381.1 | - | 3 | 542 | |||
BC031576.1 | - | 4 | 682 | 5188219 |