Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145114.1 | NP_001138586.1 | 8 | 1133 | ||
NM_001284337.1 | NP_001271266.1 | 7 | 1016 | |||
NM_015179.3 | NP_055994.2 | 10 | 1316 | |||
XM_011539555.1 | XP_011537857.1 | 9 | 1185 | |||
XM_011539556.2 | XP_011537858.1 | 10 | 1286 | |||
GenBank mRNA | AB014590.1 | - | 7 | 929 | ||
AK000238.1 | - | 7 | 726 | |||
AK022496.1 | - | 10 | 1287 | |||
AK131283.1 | - | 4 | 392 | |||
AK296315.1 | - | 8 | 997 | |||
BC012745.2 | - | 10 | 1254 | 4025242 |