Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bean1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141922.1 | NP_001135394.1 | 6 | 1206 | ||
NM_001141924.1 | NP_001135396.1 | 4 | 989 | |||
NM_001141925.1 | NP_001135397.1 | 4 | 991 | |||
GenBank mRNA | AF240460.1 | - | 2 | 386 | ||
AK134633.1 | - | 4 | 991 | |||
BC030426.1 | - | 4 | 1069 | |||
BC138184.1 | - | 6 | 1135 | 8861203 |