Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016911.4 | NP_058607.1 | 7 | 932 | ||
GenBank mRNA | AB028049.1 | - | 7 | 859 | ||
AB028050.1 | - | 5 | 607 | |||
AK034696.1 | - | 7 | 934 | |||
AK159662.1 | - | 6 | 920 | |||
BC022572.1 | - | 7 | 888 | 3993025 |