Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081069.1 | NP_001074538.1 | 16 | 1236 | ||
GenBank mRNA | AF061934.1 | - | 15 | 1141 | ||
AK039314.1 | - | 15 | 1858 | |||
AK080707.1 | - | 16 | 1330 | |||
AK161551.1 | - | 12 | 1295 | |||
AK166716.1 | - | 2 | 481 | |||
BC019741.1 | - | 5 | 568 | |||
BC049910.1 | - | 14 | 1940 |