Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122963.1 | NP_001116435.1 | 10 | 2435 | ||
NM_028487.4 | NP_082763.3 | 11 | 2495 | |||
GenBank mRNA | AK135728.1 | - | 11 | 2426 | ||
AK136894.1 | - | 11 | 2275 | |||
AK145318.1 | - | 10 | 2424 | |||
AK151283.1 | - | 9 | 1656 | |||
AK163370.1 | - | 11 | 2343 | |||
AK163390.1 | - | 11 | 2497 | |||
AK169406.1 | - | 11 | 2290 | |||
BC016083.1 | - | 9 | 1289 | 4501399 | ||
BC067075.1 | - | 10 | 2209 | 30614988 | ||
BC076635.1 | - | 10 | 2265 | |||
BC094888.1 | - | 10 | 2228 | 30628044 |