Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016780.2 | NP_058060.2 | 10 | 1519 | ||
GenBank mRNA | AF026509.1 | - | 10 | 1463 | ||
AK156518.1 | - | 6 | 776 | |||
AK157958.1 | - | 10 | 1465 | |||
BC125518.1 | - | 10 | 1519 | 40130033 | ||
BC125520.1 | - | 10 | 1519 | 40130035 |