Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111111.1 | NP_001104581.1 | 9 | 1006 | ||
GenBank mRNA | AB476647.1 | - | 8 | 882 | ||
AB476648.1 | - | 9 | 945 | |||
AK010773.1 | - | 8 | 927 | |||
AK131109.1 | - | 776 | ||||
AK155125.1 | - | 9 | 996 | |||
AK157410.1 | - | 9 | 985 | |||
AK158186.1 | - | 9 | 946 | |||
BC060054.1 | - | 9 | 1169 |