Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Whsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081102.2 | NP_001074571.2 | 1 | 543 | ||
NM_001177884.1 | NP_001171355.1 | 1 | 543 | |||
NM_175231.2 | NP_780440.2 | 1 | 543 | |||
GenBank mRNA | AK038286.1 | - | 3 | 750 | ||
BC079668.1 | - | 1 | 459 | |||
BC158094.1 | - | 2 | 617 | |||
CF532498.1 | - | 1 | 80 | |||
CF742194.1 | - | 2 | 592 |