Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194063.3 | NP_918952.2 | 3 | 284 | ||
GenBank mRNA | DQ058640.1 | - | 3 | 284 |
Gene Symbol: Rhox3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102457.4 | NP_001095927.3 | 1 | 64 |
Gene Symbol: Rhox3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184969.1 | NP_001171898.1 | 64 |