Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025245.1 | NP_001020416.1 | 4 | 826 | ||
NM_001025251.2 | NP_001020422.1 | 1 | 247 | |||
NM_001025254.2 | NP_001020425.1 | 1 | 202 | |||
NM_001025255.2 | NP_001020426.1 | 1 | 247 | |||
NM_001025256.2 | NP_001020427.1 | 1 | 247 | |||
NM_001025258.2 | NP_001020429.1 | 1 | 247 | |||
NM_001025259.2 | NP_001020430.1 | 1 | 247 | |||
NM_010777.3 | NP_034907.1 | 4 | 826 | |||
GenBank mRNA | AK005129.1 | - | 1 | 181 | ||
AK040716.1 | - | 4 | 725 | |||
AK079621.1 | - | 1 | 203 | |||
AK147966.1 | - | 1 | 195 | |||
BB633793.1 | - | 1 | 203 | |||
BC004704.1 | - | 1 | 164 | 3499588 | ||
BC051094.1 | - | 4 | 757 | 3982852 | ||
BU562877.1 | - | 185 | ||||
BY245551.1 | - | 1 | 247 | |||
CK623660.1 | - | 1 | 168 | |||
CO425201.1 | - | 1 | 169 | |||
DQ887821.1 | - | 1 | 130 | |||
L07507.1 | - | 4 | 786 | |||
L07508.1 | - | 4 | 803 | |||
M15060.1 | - | 1 | 167 | |||
M15062.1 | - | 1 | 105 | |||
X67319.1 | - | 4 | 698 |