Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031197.2 | NP_112474.2 | 7 | 928 | ||
GenBank mRNA | AK005068.1 | - | 7 | 929 | ||
AK165430.1 | - | 7 | 863 | |||
BC034675.1 | - | 7 | 847 | 4236331 | ||
X15684.1 | - | 7 | 877 | |||
X16986.1 | - | 7 | 867 |