Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaset2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083938.2 | NP_001077407.1 | 8 | 652 | ||
GenBank mRNA | AK014754.1 | - | 5 | 497 | ||
AK018722.1 | - | 8 | 652 | |||
AK019234.1 | - | 8 | 644 | |||
BC031496.1 | - | 8 | 632 | 3156549 | ||
BC089534.1 | - | 8 | 961 | 6703307 |
Gene Symbol: Rnaset2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026611.2 | NP_080887.1 | 8 | 696 | ||
GenBank mRNA | AK029149.1 | - | 650 | |||
AK084454.1 | - | 656 | ||||
AK200941.1 | - | 1 | 44 | |||
BC100330.1 | - | 8 | 621 | 30527517 |