Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027511.1 | NP_081787.1 | 1 | 357 | ||
GenBank mRNA | AK014425.1 | - | 358 |