Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134644.1 | NP_001128116.1 | 7 | 818 |
Gene Symbol: Mup11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164526.1 | NP_001157998.1 | 851 | |||
GenBank mRNA | AK013972.1 | - | 7 | 828 |
Gene Symbol: Mup16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199936.1 | NP_001186865.1 | 7 | 819 |
Gene Symbol: Mup17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200006.1 | NP_001186935.1 | 7 | 825 |