Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam208a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114879.1 | NP_001108351.1 | 14 | 1950 | ||
NM_028945.1 | NP_083221.1 | 14 | 1950 | |||
GenBank mRNA | AK044563.1 | - | 14 | 1951 |