Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010397.4 | NP_034527.1 | 5 | 1225 | ||
GenBank mRNA | AB359229.1 | - | 5 | 970 | ||
AK133985.1 | - | 6 | 1157 | |||
M35243.1 | - | 5 | 1194 | |||
M35245.1 | - | 5 | 1225 | |||
M35247.1 | - | 5 | 1187 |
Gene Symbol: H2-T9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010399.2 | NP_034529.1 | 1106 | |||
GenBank mRNA | BC044208.1 | - | 1117 | 5036676 | ||
M35248.1 | - | 1106 |