Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h3i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178207.2 | NP_835514.1 | 1 | 343 | ||
GenBank mRNA | AK155722.1 | - | 344 | |||
BC107287.2 | - | 1 | 312 | |||
BC116383.2 | - | 1 | 316 | 40101629 | ||
BC127148.1 | - | 1 | 315 |
Gene Symbol: Hist1h3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013550.5 | NP_038578.2 | 1 | 349 | ||
GenBank mRNA | AK006742.1 | - | 1 | 329 | ||
AK018952.1 | - | 1 | 350 | |||
BC115816.1 | - | 1 | 346 | 40105082 |
Gene Symbol: Hist1h3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178203.2 | NP_835510.1 | 1 | 316 |
Gene Symbol: Hist1h3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175653.2 | NP_783584.1 | 1 | 340 | ||
GenBank mRNA | BC120800.1 | - | 1 | 421 | 40129723 | |
BC120802.1 | - | 1 | 421 | 40129725 |
Gene Symbol: Hist1h3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178204.2 | NP_835511.1 | 1 | 355 | ||
GenBank mRNA | AK020421.1 | - | 356 | |||
Z30939.1 | - | 1 | 82 |
Gene Symbol: Hist1h3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178205.2 | NP_835512.1 | 1 | 316 | ||
GenBank mRNA | BC127042.1 | - | 1 | 309 |
Gene Symbol: Hist1h3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013548.4 | NP_038576.1 | 1 | 749 | ||
GenBank mRNA | AK010121.1 | - | 1 | 338 | ||
BC101954.1 | - | 1 | 749 | 40044770 | ||
BC101955.1 | - | 1 | 749 | 40044769 | ||
BC101956.1 | - | 1 | 749 | 40044765 | ||
BC103549.1 | - | 1 | 749 | 40044771 |
Gene Symbol: Hist1h3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145073.2 | NP_659539.1 | 1 | 344 | ||
GenBank mRNA | BC125355.1 | - | 1 | 340 | 40129870 | |
BC125357.1 | - | 1 | 340 | 40129872 |
Gene Symbol: Hist1h3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178206.2 | NP_835513.1 | 1 | 347 | ||
GenBank mRNA | BC107285.2 | - | 1 | 314 |