Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPIP5K2 SIRNA Assays ›
Gene Symbol:
PPIP5K2
Gene Name:
diphosphoinositol pentakisphosphate kinase 2
Gene Aliases:
CFAP160, HISPPD1, IP7K2, VIP2
Chromosome Location:
Chr.5: 103120254 - 103203523 on Build GRCh38
UniGene ID:
Hs.212046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23423
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPIP5K2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276277.1
NP_001263206.1
22
3065
NM_001281471.1
NP_001268400.1
23
3110
NM_015216.3
NP_056031.2
21
2830
XM_005271935.1
XP_005271992.1
22
3064
XM_005271936.1
XP_005271993.1
22
3064
XM_005271938.1
XP_005271995.1
22
3064
XM_005277534.1
XP_005277591.1
23
3109
XM_005277535.1
XP_005277592.1
23
3106
XM_005277536.1
XP_005277593.1
23
3109
XM_005277538.1
XP_005277595.1
23
3109
XM_005277539.1
XP_005277596.1
23
3109
XM_005277541.1
XP_005277598.1
23
3109
XM_006714576.1
XP_006714639.1
23
3109
XM_006714577.1
XP_006714640.1
23
3109
XM_011543290.2
XP_011541592.1
23
3109
XM_017009269.1
XP_016864758.1
22
3061
XM_017009270.1
XP_016864759.1
22
3064
XM_017009271.1
XP_016864760.1
22
3061
XM_017009272.1
XP_016864761.1
22
3064
XM_017009273.1
XP_016864762.1
22
3061
XM_017009274.1
XP_016864763.1
22
3064
XM_017009275.1
XP_016864764.1
22
3061
GenBank mRNA
AB007893.1
-
22
3055
AK025159.1
-
4
368
AK294787.1
-
23
2974
BC024591.1
-
21
2583
3913521
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
inositol metabolic process
phosphorylation
dephosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
acid phosphatase activity
ATP binding
diphosphoinositol-pentakisphosphate kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0