Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199079.1 | NP_951061.1 | 11 | 1678 | ||
GenBank mRNA | AK039888.1 | - | 11 | 1677 | ||
AK161001.1 | - | 11 | 1677 | |||
BC062910.1 | - | 11 | 1664 | 6816663 |