Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146023.1 | NP_001139495.1 | 4 | 397 | ||
GenBank mRNA | AK003496.1 | - | 3 | 324 | ||
AK080790.1 | - | 3 | 489 | |||
AK138395.1 | - | 4 | 371 | |||
AK166139.1 | - | 4 | 367 | |||
AK171108.1 | - | 2 | 763 | |||
BC058765.1 | - | 3 | 454 |