Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039689.1 | NP_001034778.1 | 1 | 76 | ||
GenBank mRNA | AJ972667.1 | - | 1 | 76 |
Gene Symbol: Rhox4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039698.1 | NP_001034787.1 | 1 | 151 | ||
GenBank mRNA | AJ972671.1 | - | 1 | 151 |