Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027147.1 | NP_081423.1 | 2 | 1162 | ||
GenBank mRNA | AK009710.1 | - | 2 | 1162 | ||
AK050496.1 | - | 2 | 961 | |||
AK182884.1 | - | 1 | 309 | |||
AK205625.1 | - | 1 | 309 | |||
AK206064.1 | - | 1 | 308 | |||
AK206108.1 | - | 1 | 309 | |||
AK207085.1 | - | 1 | 294 | |||
AK208424.1 | - | 1 | 309 | |||
AK214699.1 | - | 1 | 309 | |||
AK216096.1 | - | 1 | 309 | |||
AK219775.1 | - | 1 | 303 | |||
BC021944.1 | - | 2 | 820 |