Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018853.3 | NP_061341.1 | 2 | 170 | ||
GenBank mRNA | AK007832.1 | - | 2 | 201 | ||
AK010656.1 | - | 2 | 202 | |||
AK088070.1 | - | 2 | 200 | |||
AK169347.1 | - | 2 | 200 | |||
BC013252.1 | - | 2 | 135 | |||
BC058685.1 | - | 2 | 212 | 6410847 | ||
BC091747.1 | - | 2 | 175 | 2655117 | ||
BC092088.1 | - | 2 | 174 | 5102711 | ||
BC092536.1 | - | 170 | 6774435 | |||
U29402.1 | - | 2 | 198 |
Gene Symbol: Gm5244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157910.1 | - | 1 | 184 | ||
BC158067.1 | - | 1 | 275 | |||
BC172075.1 | - | 1 | 184 |