Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178193.2 | NP_835500.1 | 1 | 137 | ||
GenBank mRNA | BC132186.1 | - | 1 | 137 | 40130463 | |
BC132212.1 | - | 1 | 137 | 40130489 |