Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039050.1 | NP_001034139.1 | 5 | 1159 | ||
NM_001039051.2 | NP_001034140.1 | 5 | 965 | |||
NM_001039052.1 | NP_001034141.1 | 5 | 825 | |||
NM_001039053.2 | NP_001034142.2 | 6 | 861 | |||
NM_001190402.1 | NP_001177331.1 | 5 | 1063 | |||
NM_008863.4 | NP_032889.3 | 6 | 1073 | |||
GenBank mRNA | AK005281.1 | - | 5 | 897 | ||
AK083875.1 | - | 6 | 972 | |||
AK156950.1 | - | 5 | 852 | |||
AK157521.1 | - | 6 | 821 | |||
BC052351.1 | - | 5 | 1159 | |||
L02241.1 | - | 6 | 1074 |