Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033602.2 | NP_001028774.2 | 9, 10 | 3595 | ||
NM_015233.5 | NP_056048.1 | 4, 5 | 647 | |||
XM_011535019.2 | XP_011533321.1 | 12, 13 | 4180 | |||
XM_011535020.2 | XP_011533322.1 | 11, 12 | 4120 | |||
XM_011535021.2 | XP_011533323.1 | 11, 12 | 4117 | |||
XM_011535022.2 | XP_011533324.1 | 9, 10 | 1245 | |||
XM_017020502.1 | XP_016875991.1 | 12, 13 | 4128 | |||
GenBank mRNA | AB018317.2 | - | 9, 10 | 3595 | ||
AK293970.1 | - | 3, 4 | 559 | |||
AK302516.1 | - | 3, 4 | 338 | |||
BC032481.1 | - | 4, 5 | 647 | |||
BC119654.1 | - | 4, 5 | 578 | |||
BC119655.1 | - | 3, 4 | 511 | |||
BC129985.1 | - | 4, 5 | 578 | |||
BC150245.1 | - | 9, 10 | 3616 | 9007469 | ||
BM906724.1 | - | 4, 5 | 670 |