Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108808.1 | NP_001102278.1 | 4, 5 | 948 | ||
GenBank mRNA | BC158817.1 | - | 555 | 7374173 | ||
FQ219916.1 | - | 628 | ||||
FQ220431.1 | - | 671 | ||||
FQ226325.1 | - | 640 | ||||
FQ231706.1 | - | 640 | ||||
FQ232529.1 | - | 640 | ||||
FQ233479.1 | - | 576 |