Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107533.2 | NP_001101003.2 | 14, 15 | 1858 | ||
XM_006229477.2 | XP_006229539.1 | 14, 15 | 1853 |