Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lama4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751975.3 | XP_003752023.1 | 8 | 1582 | ||
GenBank mRNA | FQ221747.1 | - | 8 | 1264 |