Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015253.1 | NP_056068.1 | 6, 7 | 1340 | ||
XM_005256572.1 | XP_005256629.1 | 6, 7 | 1428 | |||
XM_005256573.1 | XP_005256630.1 | 6, 7 | 1403 | |||
GenBank mRNA | AB011095.1 | - | 5, 6 | 692 | ||
AK000243.1 | - | 4, 5 | 813 | |||
AK289603.1 | - | 6, 7 | 1399 | |||
AK309645.1 | - | 5, 6 | 792 | |||
AL833730.1 | - | 6, 7 | 1340 | |||
BC009975.2 | - | 6, 7 | 1365 | 4110708 |