Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106882.1 | NP_001100352.1 | 3 | 693 | ||
XM_006244421.2 | XP_006244483.1 | 5 | 1524 | |||
XM_006244422.2 | XP_006244484.1 | 4 | 1090 | |||
XM_006244423.2 | XP_006244485.1 | 3 | 1252 | |||
XM_008766843.1 | XP_008765065.1 | 3 | 1252 |