Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106633.1 | NP_001100103.1 | 3 | 375 | ||
XM_006237805.2 | XP_006237867.1 | 3 | 400 | |||
GenBank mRNA | FQ214529.1 | - | 3 | 362 | ||
FQ216110.1 | - | 3 | 391 | |||
FQ216137.1 | - | 3 | 341 | |||
FQ221067.1 | - | 3 | 360 |