Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1563348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114939.1 | NP_001108411.1 | 2, 3 | 364 | ||
GenBank mRNA | FQ220990.1 | - | 2, 3 | 373 |