Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260491.1 | NP_001247420.1 | 13 | 1487 | ||
NM_015266.2 | NP_056081.1 | 13 | 1439 | |||
NR_048537.1 | 12 | 1404 | ||||
NR_048538.1 | 11 | 1320 | ||||
NR_048539.1 | 11 | 1276 | ||||
NR_048540.1 | 9 | 1102 | ||||
XM_006723756.1 | XP_006723819.1 | 14 | 1400 | |||
XM_011528736.1 | XP_011527038.1 | 14 | 1448 | |||
XM_011528737.1 | XP_011527039.1 | 13 | 1321 | |||
XM_011528738.1 | XP_011527040.1 | 13 | 1331 | |||
XM_011528739.1 | XP_011527041.1 | 13 | 1325 | |||
XM_011528740.1 | XP_011527042.1 | 14 | 1448 | |||
XM_011528741.1 | XP_011527043.1 | 12 | 1232 | |||
XM_011528742.1 | XP_011527044.1 | 12 | 1221 | |||
XM_011528743.2 | XP_011527045.1 | 6 | 585 | |||
XM_011528744.2 | XP_011527046.1 | 5 | 416 | |||
XM_017027754.1 | XP_016883243.1 | 14 | 1448 | |||
XM_017027755.1 | XP_016883244.1 | 13 | 1309 | |||
XM_017027756.1 | XP_016883245.1 | 14 | 1397 | |||
GenBank mRNA | AB023156.2 | - | 13 | 1271 | ||
AK295665.1 | - | 11 | 1122 | |||
AK295803.1 | - | 12 | 1079 | |||
AK295822.1 | - | 12 | 1197 | |||
AK300318.1 | - | 13 | 1372 | |||
AK308328.1 | - | 9 | 906 | |||
AK310530.1 | - | 13 | 1241 | |||
AL833574.1 | - | 2 | 138 | |||
BC112213.1 | - | 13 | 1261 | 8327681 | ||
CR749638.1 | - | 13 | 1316 |