Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751784.3 | XP_003751832.1 | 2 | 492 | ||
XM_006254710.2 | XP_006254772.1 | 2 | 492 | |||
XM_006254711.2 | XP_006254773.1 | 2 | 492 | |||
XM_006254712.2 | XP_006254774.1 | 2 | 492 |