Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022586.1 | NP_072108.1 | 3 | 379 | ||
GenBank mRNA | AF148700.1 | - | 3 | 379 |