Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIM2 SIRNA Assays ›
Gene Symbol:
TRIM2
Gene Name:
tripartite motif containing 2
Gene Aliases:
CMT2R, RNF86
Chromosome Location:
Chr.4: 153152281 - 153339322 on Build GRCh38
UniGene ID:
Hs.435711
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23554
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130067.1
NP_001123539.1
9
1948
NM_015271.4
NP_056086.2
9
1945
XM_006714157.1
XP_006714220.1
9
1945
XM_006714158.1
XP_006714221.1
9
1910
XM_006714159.3
XP_006714222.1
9
2756
XM_006714160.1
XP_006714223.1
10
2710
XM_006714161.1
XP_006714224.1
10
2707
XM_006714162.1
XP_006714225.1
10
2427
XM_006714165.3
XP_006714228.1
7
2138
XM_011531794.1
XP_011530096.1
9
2024
XM_011531795.1
XP_011530097.1
10
2430
XM_011531796.2
XP_011530098.1
9
1960
XM_011531798.2
XP_011530100.1
9
2065
XM_017007943.1
XP_016863432.1
9
1910
XM_017007944.1
XP_016863433.1
10
2710
XM_017007945.1
XP_016863434.1
10
2707
XM_017007946.1
XP_016863435.1
9
2753
XM_017007947.1
XP_016863436.1
9
2024
XM_017007948.1
XP_016863437.1
9
1978
XM_017007949.1
XP_016863438.1
10
2427
XM_017007950.1
XP_016863439.1
10
2430
XM_017007951.1
XP_016863440.1
9
2124
XM_017007952.1
XP_016863441.1
9
2132
XM_017007953.1
XP_016863442.1
7
1476
XM_017007954.1
XP_016863443.1
10
2002
XM_017007955.1
XP_016863444.1
10
1995
GenBank mRNA
AB011089.1
-
9
1893
AF220018.1
-
9
1893
AL110234.1
-
2
205
BC005016.1
-
4
465
BC011052.1
-
9
1948
4156234
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108552.This siRNA also targets the following transcripts in Mouse: NM_001271727, NM_001271728, NM_001271726, NM_030706, NM_001271725, and others.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of neuron apoptotic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0