Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107494.1 | NP_001100964.1 | 2 | 429 | ||
XM_006228438.2 | XP_006228500.1 | 3 | 905 |