Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033074.1 | NP_001028246.1 | 1 | 75 | ||
XM_008771364.1 | XP_008769586.1 | 2 | 276 | |||
XM_008771365.1 | XP_008769587.1 | 2 | 276 | |||
GenBank mRNA | AY623754.1 | - | 1 | 75 |