Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037347.2 | NP_001032424.2 | 3 | 504 | ||
XM_006248024.2 | XP_006248086.1 | 3 | 606 | |||
XM_006248025.2 | XP_006248087.1 | 3 | 606 | |||
XM_008768558.1 | XP_008766780.1 | 3 | 606 | |||
GenBank mRNA | AY387086.1 | - | 4 | 261 | ||
U49058.1 | - | 3 | 515 |