Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323374.1 | NP_001310303.1 | 4 | 362 | ||
NM_001323375.1 | NP_001310304.1 | 4 | 394 | |||
NM_001323377.1 | NP_001310306.1 | 3 | 205 | |||
NM_001744.5 | NP_001735.1 | 3 | 510 | |||
GenBank mRNA | AK312840.1 | - | 3 | 365 | ||
BC016695.1 | - | 3 | 354 | |||
BC025687.1 | - | 3 | 352 | 5205478 | ||
BG721313.1 | - | 3 | 368 | |||
D30742.1 | - | 3 | 340 | |||
DA114495.1 | - | 3 | 520 | |||
DA396146.1 | - | 3 | 498 | |||
L17000.1 | - | 3 | 304 | |||
L24959.1 | - | 4 | 362 |