Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107755.1 | NP_001101225.1 | 2, 3 | 312 | ||
XM_008762061.1 | XP_008760283.1 | 2, 3 | 429 |