Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nadk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109678.1 | NP_001103148.1 | 12 | 1867 | ||
XM_006239513.2 | XP_006239575.1 | 12 | 1396 | |||
XM_006239514.2 | XP_006239576.1 | 12 | 1485 | |||
XM_006239515.2 | XP_006239577.1 | 11 | 1239 | |||
GenBank mRNA | BC090019.1 | - | 12 | 1463 | 7325365 |