Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 20 | 2690 | ||
XM_011525642.1 | XP_011523944.1 | 20 | 2690 | |||
XM_011525643.2 | XP_011523945.1 | 20 | 2690 | |||
XM_011525644.2 | XP_011523946.1 | 19 | 2598 | |||
XM_017025684.1 | XP_016881173.1 | 21 | 2868 | |||
GenBank mRNA | CR627458.1 | - | 9 | 914 |