Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DOCK9 SIRNA Assays ›
Gene Symbol:
DOCK9
Gene Name:
dedicator of cytokinesis 9
Gene Aliases:
ZIZ1, ZIZIMIN1
Chromosome Location:
Chr.13: 98793487 - 99086406 on Build GRCh38
UniGene ID:
Hs.596105
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23616
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DOCK9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130048.1
NP_001123520.1
3
447
NM_001130049.1
NP_001123521.1
3
330
NM_001130050.1
NP_001123522.1
3
447
NM_001318849.1
NP_001305778.1
3
330
NM_015296.2
NP_056111.1
3
330
XM_005254034.3
XP_005254091.1
3
319
XM_005254035.3
XP_005254092.1
3
319
XM_006719922.1
XP_006719985.1
3
480
XM_006719924.2
XP_006719987.1
3
319
XM_006719925.1
XP_006719988.1
3
447
XM_006719931.3
XP_006719994.1
3
275
XM_006719932.1
XP_006719995.1
3
447
XM_006719936.1
XP_006719999.1
3
480
XM_006719937.3
XP_006720000.1
3
275
XM_017020503.1
XP_016875992.1
3
320
XM_017020504.1
XP_016875993.1
3
320
XM_017020505.1
XP_016875994.1
3
320
XM_017020506.1
XP_016875995.1
3
320
XM_017020507.1
XP_016875996.1
3
320
XM_017020508.1
XP_016875997.1
3
320
XM_017020509.1
XP_016875998.1
3
447
XM_017020510.1
XP_016875999.1
3
320
XM_017020511.1
XP_016876000.1
3
320
XM_017020512.1
XP_016876001.1
3
447
XM_017020513.1
XP_016876002.1
3
320
XM_017020514.1
XP_016876003.1
3
447
XM_017020515.1
XP_016876004.1
3
447
XM_017020516.1
XP_016876005.1
3
447
XM_017020517.1
XP_016876006.1
3
419
XM_017020518.1
XP_016876007.1
3
419
XM_017020519.1
XP_016876008.1
3
447
XM_017020520.1
XP_016876009.1
3
447
XM_017020521.1
XP_016876010.1
3
480
GenBank mRNA
AB028981.2
-
3
353
AF527605.1
-
3
330
AK126492.1
-
3
447
AK127329.1
-
3
419
BC136274.1
-
3
310
9020261
BC146814.1
-
3
324
8860392
BC171753.1
-
3
310
9053254
CR936820.1
-
3
424
DB277756.1
-
3
448
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
blood coagulation
biological_process
positive regulation of GTPase activity
cell-cell adhesion
guanyl-nucleotide exchange factor activity
protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0