Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108050.2 | NP_001101520.2 | 4 | 1194 | ||
NM_001271174.1 | NP_001258103.1 | 4 | 1452 | |||
GenBank mRNA | FQ087395.1 | - | 1 | 172 | ||
FQ211523.1 | - | 3 | 1278 |