Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBA52 SIRNA Assays ›
Gene Symbol:
Uba52
Gene Name:
ubiquitin A-52 residue ribosomal protein fusion product 1
Gene Aliases:
Rps27a, Ubb
Chromosome Location:
Chr.16: 20668941 - 20671127 on Build Rnor_6.0
UniGene ID:
Rn.110618
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s236356
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Uba52
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031687.3
NP_113875.1
4
427
XM_006252934.2
XP_006252996.1
4
514
XM_006252935.1
XP_006252997.1
4
632
XM_008771145.1
XP_008769367.1
4
510
XM_008771146.1
XP_008769368.1
4
454
GenBank mRNA
BC061544.1
-
4
423
6922034
BP492585.1
-
3
312
FQ209944.1
-
4
419
FQ210541.1
-
4
416
FQ210904.1
-
4
417
FQ212001.1
-
4
421
FQ212205.1
-
4
419
FQ212291.1
-
4
417
FQ212810.1
-
4
420
FQ217153.1
-
4
428
FQ217440.1
-
4
412
FQ218121.1
-
4
413
FQ219376.1
-
4
418
FQ219803.1
-
4
419
FQ221045.1
-
4
416
FQ221118.1
-
4
417
FQ221238.1
-
4
637
FQ221621.1
-
4
413
FQ221631.1
-
4
419
FQ221755.1
-
4
417
FQ221772.1
-
4
417
FQ221777.1
-
4
417
FQ222045.1
-
4
417
FQ222098.1
-
4
413
FQ222371.1
-
4
416
FQ222440.1
-
4
428
FQ222483.1
-
4
416
FQ222507.1
-
4
411
FQ222700.1
-
4
416
FQ223258.1
-
4
419
FQ223423.1
-
4
413
FQ223456.1
-
4
414
FQ224647.1
-
3
407
FQ224702.1
-
4
417
FQ228362.1
-
4
413
FQ228566.1
-
4
417
U25064.1
-
1
150
X82636.1
-
3
389
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
translation
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0