Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015305.3 | NP_056120.2 | 5 | 1189 | ||
XM_017021115.1 | XP_016876604.1 | 5 | 1196 | |||
GenBank mRNA | AB018302.1 | - | 5 | 1086 | ||
AL137268.1 | - | 5 | 1087 | |||
BC028714.2 | - | 5 | 1136 | 4830897 |